Assay Details
| Gene Symbol: ctss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200426.1 | NP_956720.1 | 5 - 6 | 643 | 68 | |
| XM_017352203.2 | XP_017207692.1 | 4 - 5 | 683 | 68 | ||
| XM_017352204.2 | XP_017207693.1 | 5 - 6 | 1106 | 68 | ||
| GenBank mRNA | BC054668.1 | - | 5 - 6 | 643 | 68 | |
| BC066625.1 | - | 4 - 5 | 595 | 68 | ||
More Information
Set Membership: |
|

ctss1
Genomic Map