Assay Details
| Gene Symbol: pdhx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200560.2 | NP_956854.2 | 10 - 11 | 1266 | 66 | |
| XM_005170734.4 | XP_005170791.1 | 10 - 11 | 1241 | 66 | ||
| GenBank mRNA | BC056571.1 | - | 10 - 11 | 1232 | 66 | |
| BC067730.1 | - | 10 - 11 | 1240 | 66 | ||
| BC153624.1 | - | 7 - 8 | 714 | 66 | ||
| EH482395.1 | - | 1 - 2 | 604 | 66 | ||
More Information
Set Membership: |
|

pdhx
Genomic Map