Assay Details
| Gene Symbol: acsl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200649.1 | NP_956943.1 | 11 - 12 | 1709 | 101 | |
| GenBank mRNA | BC057476.1 | - | 11 - 12 | 1709 | 101 | |
| BC164569.1 | - | 10 - 11 | 1533 | 101 | ||
More Information
Set Membership: |
|

acsl4a
Genomic Map