Assay Details
| Gene Symbol: acsl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200649.1 | NP_956943.1 | 13 - 14 | 2016 | 113 | |
| GenBank mRNA | BC057476.1 | - | 13 - 14 | 2016 | 113 | |
| BC164569.1 | - | 12 - 13 | 1840 | 113 | ||
More Information
Set Membership: |
|

acsl4a
Genomic Map