Assay Details
| Gene Symbol: acsl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200649.1 | NP_956943.1 | 14 - 15 | 2172 | 102 | |
| GenBank mRNA | BC057476.1 | - | 14 - 15 | 2172 | 102 | |
| BC164569.1 | - | 13 - 14 | 1996 | 102 | ||
More Information
Set Membership: |
|

acsl4a
Genomic Map