Assay Details
| Gene Symbol: acsl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200649.1 | NP_956943.1 | 6 - 7 | 1130 | 71 | |
| GenBank mRNA | BC057476.1 | - | 6 - 7 | 1130 | 71 | |
| BC164569.1 | - | 5 - 6 | 954 | 71 | ||
More Information
Set Membership: |
|

acsl4a
Genomic Map