Assay Details
| Gene Symbol: rpl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200713.1 | NP_957007.1 | 3 - 4 | 510 | 72 | |
| XM_005167684.1 | XP_005167741.1 | 4 - 5 | 1081 | 72 | ||
| GenBank mRNA | BC059473.1 | - | 3 - 4 | 510 | 72 | |
| BC065432.1 | - | 3 - 4 | 507 | 72 | ||
More Information
Set Membership: |
|

rpl8
Genomic Map