Assay Details
| Gene Symbol: paip2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200766.1 | NP_957060.1 | 3 - 4 | 434 | 57 | |
| GenBank mRNA | BC059597.1 | - | 3 - 4 | 434 | 57 | |
More Information
Set Membership: |
|

paip2b
Genomic Map