Assay Details
| Gene Symbol: myoz2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200854.2 | NP_957148.2 | 2 - 3 | 241 | 143 | |
| XM_005170915.4 | XP_005170972.1 | 3 - 4 | 417 | 143 | ||
| GenBank mRNA | BC062840.1 | - | 2 - 3 | 245 | 143 | |
| EH442761.1 | - | 2 - 3 | 256 | 143 | ||
| EH578416.1 | - | 2 - 3 | 272 | 143 | ||
More Information
Set Membership: |
|

myoz2b
Genomic Map