Assay Details
| Gene Symbol: ncoa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201129.1 | NP_957423.1 | 7 - 8 | 794 | 64 | |
| XM_005156494.4 | XP_005156551.1 | 7 - 8 | 1070 | 64 | ||
| GenBank mRNA | BC047166.1 | - | 7 - 8 | 794 | 64 | |
| BC065638.1 | - | 7 - 8 | 769 | 64 | ||
| BC165720.1 | - | 6 - 7 | 700 | 64 | ||
More Information
Set Membership: |
|

ncoa4
Genomic Map