Assay Details
| Gene Symbol: nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309383.1 | NP_001296312.1 | 7 - 8 | 762 | 68 | |
| NM_201217.2 | NP_957511.2 | 8 - 9 | 1115 | 68 | ||
| GenBank mRNA | AY423035.1 | - | 8 - 9 | 1072 | 68 | |
| AY648814.1 | - | 8 - 9 | 1051 | 68 | ||
| BC056732.1 | - | 8 - 9 | 1038 | 68 | ||
| BC090915.1 | - | 7 - 8 | 707 | 68 | ||
| BC163910.1 | - | 8 - 9 | 1065 | 68 | ||
| BC163915.1 | - | 8 - 9 | 1065 | 68 | ||
| EH473181.1 | - | 3 - 4 | 326 | 68 | ||
More Information
Set Membership: |
|

nop56
Genomic Map