Assay Details
| Gene Symbol: atp6v1c1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201322.1 | NP_958479.1 | 12 - 13 | 1201 | 94 | |
| GenBank mRNA | BC053214.1 | - | 12 - 13 | 1201 | 94 | |
| BC164479.1 | - | 11 - 12 | 1083 | 94 | ||
More Information
Set Membership: |
|

atp6v1c1a
Genomic Map