Assay Details
| Gene Symbol: slc25a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205549.1 | NP_991112.1 | 2 - 3 | 222 | 68 | |
| XM_009307225.3 | XP_009305500.1 | 2 - 3 | 165 | 68 | ||
| GenBank mRNA | BC045977.1 | - | 2 - 3 | 222 | 68 | |
| BC065855.1 | - | 2 - 3 | 197 | 68 | ||
More Information
Set Membership: |
|

slc25a16
Genomic Map