Assay Details
| Gene Symbol: srsf7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205673.1 | NP_991236.1 | 6 - 7 | 687 | 78 | |
| XM_005165288.4 | XP_005165345.1 | 6 - 7 | 705 | 78 | ||
| XM_005165289.1 | XP_005165346.1 | 5 - 6 | 653 | 78 | ||
| XM_005165294.3 | XP_005165351.1 | 6 - 7 | 800 | 78 | ||
| GenBank mRNA | BC065586.1 | - | 6 - 7 | 687 | 78 | |
More Information
Set Membership: |
|

srsf7a
Genomic Map