Assay Details
Gene Symbol: c1ql3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205701.1 | NP_991264.1 | 1 - 2 | 842 | 72 | |
GenBank mRNA | BC065968.1 | - | 1 - 2 | 842 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
