Assay Details
| Gene Symbol: gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205723.1 | NP_991286.1 | 3 - 4 | 402 | 91 | |
| GenBank mRNA | AY398351.1 | - | 3 - 4 | 402 | 91 | |
| BC076004.1 | - | 3 - 4 | 360 | 91 | ||
More Information
Set Membership: |
|

gabarapl2
Genomic Map