Assay Details
| Gene Symbol: slc22a6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207077.2 | NP_996960.2 | 4 - 5 | 1007 | 96 | |
| GenBank mRNA | BC066519.1 | - | 4 - 5 | 1010 | 96 | |
More Information
Set Membership: |
|

slc22a6l
Genomic Map