Assay Details
| Gene Symbol: slc22a6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207077.2 | NP_996960.2 | 7 - 8 | 1405 | 69 | |
| GenBank mRNA | BC066519.1 | - | 7 - 8 | 1408 | 69 | |
More Information
Set Membership: |
|

slc22a6l
Genomic Map