Assay Details
| Gene Symbol: znf385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212793.2 | NP_997958.2 | 6 - 7 | 1148 | 73 | |
| XM_005167877.4 | XP_005167934.1 | 5 - 6 | 942 | 73 | ||
| XM_021478615.1 | XP_021334290.1 | 6 - 7 | 1202 | 73 | ||
| XM_021478616.1 | XP_021334291.1 | 5 - 6 | 917 | 73 | ||
| XM_021478617.1 | XP_021334292.1 | 5 - 6 | 989 | 73 | ||
| GenBank mRNA | BC059587.1 | - | 6 - 7 | 1151 | 73 | |
More Information
Set Membership: |
|

znf385b
Genomic Map