Assay Details
| Gene Symbol: dip2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212963.1 | NP_998128.1 | 13 - 14 | 1798 | 83 | |
| XM_009296840.3 | XP_009295115.1 | 14 - 15 | 1842 | 83 | ||
| XM_009296841.3 | XP_009295116.1 | 13 - 14 | 1836 | 83 | ||
| XM_009296842.3 | XP_009295117.1 | 14 - 15 | 1842 | 83 | ||
| GenBank mRNA | BC068015.1 | - | 13 - 14 | 1798 | 83 | |
More Information
Set Membership: |
|

dip2ba
Genomic Map