Assay Details
| Gene Symbol: dip2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212963.1 | NP_998128.1 | 14 - 15 | 1862 | 130 | |
| XM_009296840.3 | XP_009295115.1 | 15 - 16 | 1906 | 130 | ||
| XM_009296841.3 | XP_009295116.1 | 14 - 15 | 1900 | 130 | ||
| XM_009296842.3 | XP_009295117.1 | 15 - 16 | 1906 | 130 | ||
| GenBank mRNA | BC068015.1 | - | 14 - 15 | 1862 | 130 | |
More Information
Set Membership: |
|

dip2ba
Genomic Map