Assay Details
| Gene Symbol: dip2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212963.1 | NP_998128.1 | 19 - 20 | 2470 | 103 | |
| XM_009296840.3 | XP_009295115.1 | 20 - 21 | 2514 | 103 | ||
| XM_009296841.3 | XP_009295116.1 | 19 - 20 | 2508 | 103 | ||
| XM_009296842.3 | XP_009295117.1 | 20 - 21 | 2514 | 103 | ||
| GenBank mRNA | BC068015.1 | - | 19 - 20 | 2470 | 103 | |
More Information
Set Membership: |
|

dip2ba
Genomic Map