Assay Details
| Gene Symbol: dip2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212963.1 | NP_998128.1 | 29 - 30 | 3660 | 66 | |
| XM_009296840.3 | XP_009295115.1 | 31 - 32 | 3728 | 66 | ||
| XM_009296841.3 | XP_009295116.1 | 30 - 31 | 3722 | 66 | ||
| XM_009296842.3 | XP_009295117.1 | 30 - 31 | 3704 | 66 | ||
| GenBank mRNA | BC068015.1 | - | 29 - 30 | 3660 | 66 | |
More Information
Set Membership: |
|

dip2ba
Genomic Map