Assay Details
| Gene Symbol: dip2ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212963.1 | NP_998128.1 | 37 - 38 | 4623 | 65 | |
| XM_009296840.3 | XP_009295115.1 | 39 - 40 | 4691 | 65 | ||
| XM_009296841.3 | XP_009295116.1 | 38 - 39 | 4685 | 65 | ||
| XM_009296842.3 | XP_009295117.1 | 38 - 39 | 4667 | 65 | ||
| GenBank mRNA | BC068015.1 | - | 37 - 38 | 4623 | 65 | |
More Information
Set Membership: |
|

dip2ba
Genomic Map