Assay Details
| Gene Symbol: eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212990.1 | NP_998155.1 | 6 - 7 | 687 | 79 | |
| GenBank mRNA | BC045995.1 | - | 6 - 7 | 687 | 79 | |
| BC065874.1 | - | 6 - 7 | 79 | |||
More Information
Set Membership: |
|

eif3i
Genomic Map