Assay Details
| Gene Symbol: rhoaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213137.1 | NP_998302.1 | 4 - 5 | 470 | 74 | |
| GenBank mRNA | AY865555.1 | - | 3 - 4 | 415 | 74 | |
| BC056556.1 | - | 4 - 5 | 470 | 74 | ||
| BC068390.1 | - | 4 - 5 | 452 | 74 | ||
More Information
Set Membership: |
|

rhoaa
Genomic Map