Assay Details
| Gene Symbol: sh3bp5la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213160.1 | NP_998325.1 | 2 - 3 | 305 | 86 | |
| GenBank mRNA | BC057413.1 | - | 2 - 3 | 305 | 86 | |
More Information
Set Membership: |
|

sh3bp5la
Genomic Map