Assay Details
| Gene Symbol: mgst3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213427.1 | NP_998592.1 | 2 - 3 | 291 | 93 | |
| XM_005160877.2 | XP_005160934.1 | 3 - 4 | 320 | 93 | ||
| GenBank mRNA | BC049037.1 | - | 2 - 3 | 291 | 93 | |
More Information
Set Membership: |
|

mgst3a
Genomic Map