Assay Details
| Gene Symbol: pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213430.1 | NP_998595.2 | 3 - 4 | 639 | 78 | |
| XM_005156519.4 | XP_005156576.1 | 3 - 4 | 675 | 78 | ||
| GenBank mRNA | BC052113.1 | - | 3 - 4 | 639 | 78 | |
| BC065857.1 | - | 3 - 4 | 617 | 78 | ||
| BC093127.1 | - | 3 - 4 | 654 | 78 | ||
| BC155273.1 | - | 3 - 4 | 645 | 78 | ||
More Information
Set Membership: |
|

pla2g12b
Genomic Map