Assay Details
| Gene Symbol: tfdp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213492.2 | NP_998657.1 | 4 - 5 | 338 | 62 | |
| XM_005167824.4 | XP_005167881.1 | 4 - 5 | 388 | 62 | ||
| XM_009304816.2 | XP_009303091.1 | 4 - 5 | 355 | 62 | ||
| GenBank mRNA | BC044409.1 | - | 4 - 5 | 337 | 62 | |
| BC165791.1 | - | 3 - 4 | 211 | 62 | ||
More Information
Set Membership: |
|

tfdp1a
Genomic Map