Assay Details
| Gene Symbol: dnaja2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213493.1 | NP_998658.1 | 5 - 6 | 859 | 73 | |
| GenBank mRNA | BC048042.2 | - | 5 - 6 | 859 | 73 | |
| BC068384.1 | - | 5 - 6 | 600 | 73 | ||
| BC165792.1 | - | 5 - 6 | 612 | 73 | ||
More Information
Set Membership: |
|

dnaja2a
Genomic Map