Assay Details
| Gene Symbol: cnot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214760.2 | NP_999925.1 | 6 - 7 | 849 | 73 | |
| GenBank mRNA | BC067547.1 | - | 6 - 7 | 741 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cnot9| Gene Symbol: cnot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214760.2 | NP_999925.1 | 6 - 7 | 849 | 73 | |
| GenBank mRNA | BC067547.1 | - | 6 - 7 | 741 | 73 | |
Set Membership: |
|