Assay Details
| Gene Symbol: ctcf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001844.1 | NP_001001844.1 | 2 - 3 | 696 | 57 | |
| GenBank mRNA | AY547282.1 | - | 2 - 3 | 696 | 57 | |
| BC083236.1 | - | 2 - 3 | 719 | 57 | ||
| BC097009.1 | - | 2 - 3 | 710 | 57 | ||
| BC115148.1 | - | 2 - 3 | 710 | 57 | ||
| BC121782.1 | - | 2 - 3 | 715 | 57 | ||
| BC129307.1 | - | 2 - 3 | 716 | 57 | ||
| BC163931.1 | - | 2 - 3 | 603 | 57 | ||
| BC163935.1 | - | 2 - 3 | 603 | 57 | ||
More Information
Set Membership: |
|

ctcf
Genomic Map