Assay Details
| Gene Symbol: trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001849.1 | NP_001001849.1 | 5 - 6 | 595 | 63 | |
| XM_005173468.2 | XP_005173525.1 | 5 - 6 | 603 | 63 | ||
| XM_005173469.3 | XP_005173526.1 | 5 - 6 | 615 | 63 | ||
| GenBank mRNA | AY325807.1 | - | 5 - 6 | 595 | 63 | |
| BC093109.1 | - | 5 - 6 | 608 | 63 | ||
| BC121781.1 | - | 5 - 6 | 607 | 63 | ||
| BC139599.1 | - | 5 - 6 | 594 | 63 | ||
| HQ676591.1 | - | 4 - 5 | 497 | 63 | ||
More Information
Set Membership: |
|

trpv6
Genomic Map