Assay Details
| Gene Symbol: cbx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002090.1 | NP_001002090.1 | 2 - 3 | 179 | 82 | |
| XM_005156237.4 | XP_005156294.1 | 2 - 3 | 561 | 82 | ||
| XM_005156239.4 | XP_005156296.1 | 2 - 3 | 216 | 82 | ||
| XM_005156240.4 | XP_005156297.1 | 2 - 3 | 559 | 82 | ||
| GenBank mRNA | BC071539.1 | - | 2 - 3 | 179 | 82 | |
| BC097012.1 | - | 2 - 3 | 558 | 82 | ||
More Information
Set Membership: |
|

cbx1b
Genomic Map