Assay Details
| Gene Symbol: dnajc5aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002464.1 | NP_001002464.1 | 4 - 5 | 653 | 59 | |
| GenBank mRNA | BC076354.1 | - | 4 - 5 | 653 | 59 | |
| BC165285.1 | - | 3 - 4 | 536 | 59 | ||
More Information
Set Membership: |
|

dnajc5aa
Genomic Map