Assay Details
| Gene Symbol: jdp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002493.1 | NP_001002493.1 | 2 - 3 | 316 | 70 | |
| XM_005160857.3 | XP_005160914.1 | 2 - 3 | 316 | 70 | ||
| GenBank mRNA | BC076313.1 | - | 2 - 3 | 316 | 70 | |
More Information
Set Membership: |
|

jdp2b
Genomic Map