Assay Details
| Gene Symbol: thap12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002680.1 | NP_001002680.1 | 1 - 2 | 159 | 58 | |
| GenBank mRNA | BC075781.1 | - | 1 - 2 | 159 | 58 | |
More Information
Set Membership: |
|

thap12b
Genomic Map