Assay Details
| Gene Symbol: rasgef1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003479.1 | NP_001003479.1 | 7 - 8 | 1069 | 60 | |
| GenBank mRNA | BC078340.1 | - | 7 - 8 | 1069 | 60 | |
More Information
Set Membership: |
|

rasgef1bb
Genomic Map