Assay Details
| Gene Symbol: rasgef1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003479.1 | NP_001003479.1 | 8 - 9 | 1179 | 85 | |
| GenBank mRNA | BC078340.1 | - | 8 - 9 | 1179 | 85 | |
More Information
Set Membership: |
|

rasgef1bb
Genomic Map