Assay Details
| Gene Symbol: tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003550.2 | NP_001003550.1 | 8 - 9 | 1587 | 64 | |
| XM_021470138.1 | XP_021325813.1 | 8 - 9 | 1844 | 64 | ||
| GenBank mRNA | BC078291.1 | - | 8 - 9 | 1591 | 64 | |
More Information
Set Membership: |
|

tm9sf1
Genomic Map