Assay Details
| Gene Symbol: aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003749.4 | NP_001003749.1 | 2 - 3 | 480 | 61 | |
| NM_001358313.1 | NP_001345242.1 | 2 - 3 | 480 | 61 | ||
| XM_009294620.3 | XP_009292895.1 | 2 - 3 | 638 | 61 | ||
| GenBank mRNA | BC078213.1 | - | 2 - 3 | 467 | 61 | |
| CK682060.1 | - | 2 - 3 | 498 | 61 | ||
| FJ666327.1 | - | 2 - 3 | 467 | 61 | ||
More Information
Set Membership: |
|

aqp4
Genomic Map