Assay Details
| Gene Symbol: pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004117.3 | NP_001004117.2 | 5 - 6 | 698 | 99 | |
| GenBank mRNA | BC080227.1 | - | 5 - 6 | 686 | 99 | |
More Information
Set Membership: |
|

pgls
Genomic Map