Assay Details
| Gene Symbol: tmem267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003574.1 | NP_001003574.1 | 2 - 3 | 487 | 61 | |
| NM_001004514.1 | NP_001004514.1 | 1 - 2 | 371 | 61 | ||
| GenBank mRNA | BC078204.1 | - | 2 - 3 | 487 | 61 | |
| BC107628.1 | - | 2 - 3 | 471 | 61 | ||
More Information
Set Membership: |
|

tmem267
Genomic Map