Assay Details
| Gene Symbol: ppifa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004626.1 | NP_001004626.1 | 2 - 3 | 268 | 101 | |
| GenBank mRNA | BC081399.1 | - | 2 - 3 | 268 | 101 | |
| BC164981.1 | - | 2 - 3 | 208 | 101 | ||
More Information
Set Membership: |
|

ppifa
Genomic Map