Assay Details
| Gene Symbol: eif1axa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006082.2 | NP_001006082.1 | 2 - 3 | 273 | 68 | |
| GenBank mRNA | BC083238.1 | - | 2 - 3 | 275 | 68 | |
| BC164226.1 | - | 2 - 3 | 140 | 68 | ||
More Information
Set Membership: |
|

eif1axa
Genomic Map