Assay Details
| Gene Symbol: mxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007284.2 | NP_001007285.1 | 2 - 3 | 366 | 69 | |
| XM_005167671.4 | XP_005167728.1 | 3 - 4 | 561 | 69 | ||
| GenBank mRNA | BC165716.1 | - | 2 - 3 | 388 | 69 | |
| CK143042.1 | - | 2 - 3 | 374 | 69 | ||
More Information
Set Membership: |
|

mxc
Genomic Map