Assay Details
| Gene Symbol: mxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007284.2 | NP_001007285.1 | 6 - 7 | 939 | 143 | |
| XM_005167671.4 | XP_005167728.1 | 7 - 8 | 1134 | 143 | ||
| GenBank mRNA | BC165716.1 | - | 6 - 7 | 961 | 143 | |
More Information
Set Membership: |
|

mxc
Genomic Map