Assay Details
| Gene Symbol: mxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007284.2 | NP_001007285.1 | 7 - 8 | 1066 | 91 | |
| XM_005167671.4 | XP_005167728.1 | 8 - 9 | 1261 | 91 | ||
| GenBank mRNA | BC165716.1 | - | 7 - 8 | 1088 | 91 | |
More Information
Set Membership: |
|

mxc
Genomic Map