Assay Details
| Gene Symbol: mxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007284.2 | NP_001007285.1 | 9 - 10 | 1353 | 69 | |
| XM_005167671.4 | XP_005167728.1 | 10 - 11 | 1548 | 69 | ||
| GenBank mRNA | BC165716.1 | - | 9 - 10 | 1375 | 69 | |
More Information
Set Membership: |
|

mxc
Genomic Map